Learn More
Medchemexpress LLC BIP-135 | 941575-71-9 | 99.9% | 421.24 | 10 MG
SDP
Catalog No. 5000343996
Encompass_Preferred
BIP-135 is a potent and selective ATP-competitive GSK-3 inhibitor, with IC50s of 16 nM and 21 nM for GSK-3α and GSK-3β, respectively. It exhibits neuroprotective effects.
  • Potent and selective ATP-competitive GSK-3 inhibitor.
  • Exhibits neuroprotective effect.
  • Increases survival motor neuron (SMN) protein levels in human SMA fibroblasts (at 25 μM).
  • Superior neuroprotective agent in models of oxidative stress (at 20 μM).
  • Prolongs median survival time in a Δ7 SMA KO mouse model of spinal muscular atrophy (at 75 mg/kg; i.p.; daily).
Catalog No. 50-003-43996 Supplier Medchemexpress LLC Supplier No. HY11105510MG
BIP-135 is a potent and selective ATP-competitive GSK-3 inhibitor, with IC50s of 16 nM and 21 nM for GSK-3α and GSK-3β, respectively. It exhibits neuroprotective effects.
  • Potent and selective ATP-competitive GSK-3 inhibitor.
  • Exhibits neuroprotective effect.
  • Increases survival motor neuron (SMN) protein levels in human SMA fibroblasts (at 25 μM).
  • Superior neuroprotective agent in models of oxidative stress (at 20 μM).
  • Prolongs median survival time in a Δ7 SMA KO mouse model of spinal muscular atrophy (at 75 mg/kg; i.p.; daily).