Learn More
Selleck Chemical LLC BiP Inducer X -E0362-5MG
SDP
Supplier:  Selleck Chemical LLC E03625MG
Encompass_Preferred
BiP Inducer X is a selective GRP78 (BiP HSPA5) inducer BIX induces BiP only in a dose-dependent manner without induction of other molecules involved in the ER stress response
Catalog No. 50-238-9996