Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biperiden | 514-65-8 | 99.7% | 311.46 g/mol | C21H29NO | 5 MG
SDP

Catalog No. 5000262850
Encompass_Preferred

Biperiden is a research-grade, non-selective muscarinic acetylcholine receptor antagonist supplied as a solid for laboratory use. It is intended for pharmacology and neuroscience research where a characterized muscarinic antagonist is required.

  • High purity (99.74%).
  • Molecular formula C21H29NO, molecular weight 311.46 g/mol.
  • Physical form: solid, white to off-white.
  • Storage: powder stable at -20°C for long-term storage, 4°C for shorter-term storage.
  • Suitable for receptor pharmacology, neuroscience, and pharmacodynamic studies.

Catalog No. 50-002-62850 Supplier Medchemexpress LLC Supplier No. HY13204A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biperiden is a research-grade, non-selective muscarinic acetylcholine receptor antagonist supplied as a solid for laboratory use. It is intended for pharmacology and neuroscience research where a characterized muscarinic antagonist is required.

  • High purity (99.74%).
  • Molecular formula C21H29NO, molecular weight 311.46 g/mol.
  • Physical form: solid, white to off-white.
  • Storage: powder stable at -20°C for long-term storage, 4°C for shorter-term storage.
  • Suitable for receptor pharmacology, neuroscience, and pharmacodynamic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.