Learn More
Medchemexpress LLC Biperiden | 514-65-8 | MFCD05662335 | 99.7% | C21H29NO | 10MG
SDP
Catalog No. 5000260594
Encompass_Preferred
Biperiden is a tertiary amine antimuscarinic used in neuroscience and pharmacology research as a muscarinic receptor antagonist. The compound has molecular formula C21H29NO (MW ~311.5 g/mol) and CAS 514-65-8. Supplied as a research-grade sample with reported purity around 99.7%. For research use only; not for human consumption.
  • Non-selective muscarinic receptor antagonist suitable for receptor pharmacology studies.
  • High reported purity (~99.7%) supports analytical and biochemical assays.
  • Well-characterized chemical identity with established database entries for reference.
  • Compatible with common organic solvents for formulation and experimental dosing.
Catalog No. 50-002-60594 Supplier Medchemexpress LLC Supplier No. HY13204A10MG
Biperiden is a tertiary amine antimuscarinic used in neuroscience and pharmacology research as a muscarinic receptor antagonist. The compound has molecular formula C21H29NO (MW ~311.5 g/mol) and CAS 514-65-8. Supplied as a research-grade sample with reported purity around 99.7%. For research use only; not for human consumption.
  • Non-selective muscarinic receptor antagonist suitable for receptor pharmacology studies.
  • High reported purity (~99.7%) supports analytical and biochemical assays.
  • Well-characterized chemical identity with established database entries for reference.
  • Compatible with common organic solvents for formulation and experimental dosing.