Learn More
Medchemexpress LLC Biperiden hydrochloride | 1235-82-1 | MFCD00797516 | 99.9% | C21H30ClNO | 10MG
SDP
Catalog No. 5000260947
Encompass_Preferred
Biperiden hydrochloride is the hydrochloride salt of biperiden, a non-selective muscarinic receptor antagonist commonly used in preclinical research to investigate cholinergic neurotransmission and movement disorder models. It is provided as a high-purity solid and as ready-to-use solutions for laboratory assays.
  • Non-selective muscarinic receptor antagonist useful for cholinergic signaling studies.
  • High purity suitable for analytical and preclinical applications.
  • Available as solid powder and as ready-to-use DMSO solution.
  • Stable under recommended storage conditions for reliable results.
  • Soluble in common organic solvents for assay preparation.
Catalog No. 50-002-60947 Supplier Medchemexpress LLC Supplier No. HY1320410MG
Biperiden hydrochloride is the hydrochloride salt of biperiden, a non-selective muscarinic receptor antagonist commonly used in preclinical research to investigate cholinergic neurotransmission and movement disorder models. It is provided as a high-purity solid and as ready-to-use solutions for laboratory assays.
  • Non-selective muscarinic receptor antagonist useful for cholinergic signaling studies.
  • High purity suitable for analytical and preclinical applications.
  • Available as solid powder and as ready-to-use DMSO solution.
  • Stable under recommended storage conditions for reliable results.
  • Soluble in common organic solvents for assay preparation.