Learn More
Medchemexpress LLC Biphenyl-3,3,4,4-tetrol | 3598-30-9 | MFCD00038783 | 98.3% | 218.20 g/mol | C12H10O4 | 250 MG
SDP
Catalog No. 5000284056
Encompass_Preferred
Biphenyl-3′,3,4,4′-tetrol (BPT) is a small-molecule research compound reported as a potent Aβ40 aggregation inhibitor used to investigate amyloid-β aggregation in neurodegenerative disease studies. It is supplied as a high-purity solid suitable for in vitro biochemical assays.
  • Potent Aβ40 aggregation inhibitor for amyloid-β research.
  • High purity (98.3%) suitable for biochemical assays.
  • Low molecular weight (218.20 g/mol) and formula C12H10O4 for straightforward characterization.
  • Solid form, convenient for handling, storage, and formulation.
  • Standard identifiers (CAS 3598-30-9, MFCD00038783) to aid data lookup and registration.
Catalog No. 50-002-84056 Supplier Medchemexpress LLC Supplier No. HYW594851250MG
Biphenyl-3′,3,4,4′-tetrol (BPT) is a small-molecule research compound reported as a potent Aβ40 aggregation inhibitor used to investigate amyloid-β aggregation in neurodegenerative disease studies. It is supplied as a high-purity solid suitable for in vitro biochemical assays.
  • Potent Aβ40 aggregation inhibitor for amyloid-β research.
  • High purity (98.3%) suitable for biochemical assays.
  • Low molecular weight (218.20 g/mol) and formula C12H10O4 for straightforward characterization.
  • Solid form, convenient for handling, storage, and formulation.
  • Standard identifiers (CAS 3598-30-9, MFCD00038783) to aid data lookup and registration.