Learn More
Medchemexpress LLC Biphenyl-3′,3,4,4′-tetrol | 3598-30-9 | MFCD00038783 | 98.3% | 218.21 g·mol-1 | C12H10O4 | 500 MG
SDP
Catalog No. 5000284277
Encompass_Preferred
Biphenyl-3′,3,4,4′-tetrol (BPT) is a small-molecule research reagent that inhibits Aβ40 peptide aggregation and is used in studies of neurodegenerative diseases such as Alzheimer's disease. The compound is supplied as a solid with high purity and is available in multiple pack sizes, including a 500 mg package.
  • Potent Aβ40 aggregation inhibitor suitable for neurodegeneration research.
  • High purity: 98.3% (as supplied).
  • Molecular weight 218.21 g·mol-1 and formula C12H10O4.
  • Supplied as a solid in multiple pack sizes, including 500 mg.
  • Provided for research use only; follow appropriate SDS handling and storage instructions.
Catalog No. 50-002-84277 Supplier Medchemexpress LLC Supplier No. HYW594851500MG
Biphenyl-3′,3,4,4′-tetrol (BPT) is a small-molecule research reagent that inhibits Aβ40 peptide aggregation and is used in studies of neurodegenerative diseases such as Alzheimer's disease. The compound is supplied as a solid with high purity and is available in multiple pack sizes, including a 500 mg package.
  • Potent Aβ40 aggregation inhibitor suitable for neurodegeneration research.
  • High purity: 98.3% (as supplied).
  • Molecular weight 218.21 g·mol-1 and formula C12H10O4.
  • Supplied as a solid in multiple pack sizes, including 500 mg.
  • Provided for research use only; follow appropriate SDS handling and storage instructions.