Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenyl-4,4'-diboronic acid, 94%

Catalog No. aal1332803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1332803 1 g
AAL1332806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1332803 Supplier Thermo Scientific Chemicals Supplier No. L1332803
Only null left
Add to Cart

CAS: 4151-80-8 | C12H12B2O4 | 241.84 g/mol

Biphenyl-4,4'-diboronic acid is used in the preparation of cycloparaphenylenes through Suzuki coupling. It is also employed in the synthesis of 1,3,2-diazaboroine derivatives, which is useful for organic thin-film transistor applications. It plays an important role in the preparation of arylboronates from reactions with catechol, dihydroxynaphthalene and diaminobenzenedithiol, which finds application as organic field-effect transistors and light emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4,4′-diboronic acid is used in the preparation of cycloparaphenylenes through Suzuki coupling. It is also employed in the synthesis of 1,3,2-diazaboroine derivatives, which is useful for organic thin-film transistor applications. It plays an important role in the preparation of arylboronates from reactions with catechol, dihydroxynaphthalene and diaminobenzenedithiol, which finds application as organic field-effect transistors and light emitting diodes.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4151-80-8
Molecular Formula C12H12B2O4
Molecular Weight (g/mol) 241.84
MDL Number MFCD00151795
InChI Key SLHKDOGTVUCXKX-UHFFFAOYSA-N
Synonym 4,4'-biphenyldiboronic acid, 1,1'-biphenyl-4,4'-diyldiboronic acid, biphenyl-4,4'-diboronic acid, 4-4-boronophenyl phenyl boronic acid, biphenyl-4,4'-diyldiboronic acid, pubchem5324, acmc-1apw8, 4,4'-biphenyldiboric acid, 4,4-biphenyldiboronic acid, 4,4'-diboronobiphenyl
PubChem CID 2734608
IUPAC Name [4-(4-boronophenyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point ∽300°C (decomposition)
Quantity 1 g
Beilstein 2941876
Solubility Information Soluble in water.
Formula Weight 241.85
Percent Purity 95%
Chemical Name or Material Biphenyl-4,4'-diboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.