Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biphenyl-4-amidoxime | 40019-44-1 | 99.8% | 50 MG
SDP

Catalog No. 5000338547
Encompass_Preferred

Biphenyl-4-amidoxime (N'-hydroxy-4-phenylbenzene-1-carboximidamide) is a synthetic intermediate useful for pharmaceutical synthesis. It is presented as a solid, white to off-white in color, and is strictly for research use only.

  • Purity: 99.8%
  • Molecular weight: 212.25
  • Chemical formula: C13H12N2O
  • CAS number: 40019-44-1
  • Appearance: solid
  • Color: white to off-white
  • Storage as powder: -20°C for 3 years or 4°C for 2 years
  • Storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO: 87.5 mg/mL

Catalog No. 50-003-38547 Supplier Medchemexpress LLC Supplier No. HY12984450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biphenyl-4-amidoxime (N'-hydroxy-4-phenylbenzene-1-carboximidamide) is a synthetic intermediate useful for pharmaceutical synthesis. It is presented as a solid, white to off-white in color, and is strictly for research use only.

  • Purity: 99.8%
  • Molecular weight: 212.25
  • Chemical formula: C13H12N2O
  • CAS number: 40019-44-1
  • Appearance: solid
  • Color: white to off-white
  • Storage as powder: -20°C for 3 years or 4°C for 2 years
  • Storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO: 87.5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.