Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biphenyl-4-amidoxime | 40019-44-1 | 99.8% | 500 MG
SDP

Catalog No. 5000339129
Encompass_Preferred

Biphenyl-4-amidoxime, also known as N'-hydroxy-4-phenylbenzene-1-carboximidamide, is a synthetic intermediate primarily used for pharmaceutical synthesis. It presents as a white to off-white solid and has a molecular weight of 212.25 with the chemical formula C13H12N2O. This compound is intended for research use only.

  • Synthetic intermediate for pharmaceutical synthesis
  • Molecular weight of 212.25
  • Chemical formula C13H12N2O
  • Appears as a white to off-white solid
  • High purity of 99.8%
  • Intended for research use only

Catalog No. 50-003-39129 Supplier Medchemexpress LLC Supplier No. HY129844500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biphenyl-4-amidoxime, also known as N'-hydroxy-4-phenylbenzene-1-carboximidamide, is a synthetic intermediate primarily used for pharmaceutical synthesis. It presents as a white to off-white solid and has a molecular weight of 212.25 with the chemical formula C13H12N2O. This compound is intended for research use only.

  • Synthetic intermediate for pharmaceutical synthesis
  • Molecular weight of 212.25
  • Chemical formula C13H12N2O
  • Appears as a white to off-white solid
  • High purity of 99.8%
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.