Learn More
Selleck Chemical LLC Biphenyl-4-sulfonyl chloride S5438-25mg
SDP
Catalog No. 503109518
Encompass_Preferred
Biphenyl-4-sulfonyl chloride (p-Phenylbenzenesulfonyl 4-Phenylbenzenesulfonyl p-Biphenylsulfonyl) is a HDAC inhibitor with synthetic applications in palladium-catalyzed desulfitative C-arylation
Catalog No. 50-310-9518 Supplier Selleck Chemical LLC Supplier No. S543825MG
Biphenyl-4-sulfonyl chloride (p-Phenylbenzenesulfonyl 4-Phenylbenzenesulfonyl p-Biphenylsulfonyl) is a HDAC inhibitor with synthetic applications in palladium-catalyzed desulfitative C-arylation