Learn More
Medchemexpress LLC HY-15442 1mg Medchemexpress, Biphenylindanone A CAS:866823-73-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY154421MG
Encompass_Preferred
Medchemexpress, HY-15442 1mg Biphenylindanone A CAS:866823-73-6 Biphenylindanone A (BINA) is a selective human mGluR2 (hmGluR2) potentiator for the treatment of many neurological disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9940