Learn More
Cayman Chemical Bipiperidinyl 4-ANPP, Purity: ≥98%, CAS: 2515737-24-1, 5 mg
SDP
Catalog No. 502950339
Encompass_Preferred
An analytical reference standard that is structurally similar to known opioids; a byproduct in the synthesis of fentanyl; intended for research and forensic applications
Catalog No. 50-295-0339 Supplier Cayman Chemical Supplier No. 338005 MG
An analytical reference standard that is structurally similar to known opioids; a byproduct in the synthesis of fentanyl; intended for research and forensic applications