Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biprofen | 6341-72-6 | 99.7% | 226.28 g·mol⁻¹ | C15H14O2 | 250 MG
SDP

Catalog No. 5000208481
Encompass_Preferred

Biprofen is a nonsteroidal anti-inflammatory research compound supplied for laboratory and analytical use. It is provided in multiple small-scale pack sizes with reported high purity and typical storage recommendations for small-molecule reagents.

  • White to off-white solid.
  • Cas number 6341-72-6.
  • Molecular formula C15H14O2.
  • Molecular weight 226.28 g·mol⁻¹.
  • Available pack sizes include 50 mg, 100 mg, 250 mg, 500 mg, and 1 g; larger quantities by quote.
  • Purity reported around 99.7% (source shows a secondary value of 99.87%).
  • Storage: room temperature for up to 3 years; in solution, -80°C for 2 years or -20°C for 1 year.

Catalog No. 50-002-08481 Supplier Medchemexpress LLC Supplier No. 250MGHYW087195
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biprofen is a nonsteroidal anti-inflammatory research compound supplied for laboratory and analytical use. It is provided in multiple small-scale pack sizes with reported high purity and typical storage recommendations for small-molecule reagents.

  • White to off-white solid.
  • Cas number 6341-72-6.
  • Molecular formula C15H14O2.
  • Molecular weight 226.28 g·mol⁻¹.
  • Available pack sizes include 50 mg, 100 mg, 250 mg, 500 mg, and 1 g; larger quantities by quote.
  • Purity reported around 99.7% (source shows a secondary value of 99.87%).
  • Storage: room temperature for up to 3 years; in solution, -80°C for 2 years or -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.