Learn More
AVIDITY LLC BirA biotin-protein ligase bulk lyophilized reaction kit
Catalog No. NC1870426
Encompass
The BirA biotin-protein ligase adds d-biotin covalently to biotin-acceptor peptides/proteins via an ATP intermediate (biotinyl 5'-adenylate) in a highly efficient and targeted manner. This kit includes 300 ug of BirA enzyme for those who require larger scale reactions or more frequent use of the BirA biotinylation reaction in their work.
Catalog No. NC1870426 Supplier AVIDITY LLC Supplier No. BULK BIRART
The BirA biotin-protein ligase adds d-biotin covalently to biotin-acceptor peptides/proteins via an ATP intermediate (biotinyl 5'-adenylate) in a highly efficient and targeted manner. This kit includes 300 ug of BirA enzyme for those who require larger scale reactions or more frequent use of the BirA biotinylation reaction in their work.