Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVIDITY LLC BirA Biotin-Protein Ligase Kit, EC6.3.4.15, 40 μg enzyme, 1 mg/mL concentration, 2 x 1.5 mL BiomixA (0.5 M Bicine buffer, pH 8.3), 2 x 1.5 mL BiomixB (100 mM ATP, 100 mM Mg(OAc)₂, 500 μM d-biotin)

Catalog No. NC2683380
Encompass

The Avidity LLC BirA Biotin-Protein Ligase Kit provides a highly efficient method for the covalent biotinylation of peptides and proteins. Utilizing the BirA biotin-protein ligase enzyme, the kit facilitates the specific labeling of target sequences such as the AviTag™ peptide, enhancing reaction rates and minimizing potential issues with protein stability. Ideal for applications like affinity chromatography and sensitive protein detection, this kit is essential for robust labeling in research applications.

  • Product type: Biotin-protein ligase reaction kit
  • Enzyme source: E. coli (wild-type)
  • Enzyme purity: >99%
  • Required cofactor: ATP
  • Biotinylation type: Covalent
  • Standard kit enzyme concentration: 1 mg/mL
  • Standard kit enzyme amount: 40 μg
  • Main application: Protein labeling and detection
  • Key benefit: High efficiency and specificity

Catalog No. NC2683380 Supplier AVIDITY LLC Supplier No. EC6.3.4.15
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Avidity LLC BirA Biotin-Protein Ligase Kit provides a highly efficient method for the covalent biotinylation of peptides and proteins. Utilizing the BirA biotin-protein ligase enzyme, the kit facilitates the specific labeling of target sequences such as the AviTag™ peptide, enhancing reaction rates and minimizing potential issues with protein stability. Ideal for applications like affinity chromatography and sensitive protein detection, this kit is essential for robust labeling in research applications.

  • Product type: Biotin-protein ligase reaction kit
  • Enzyme source: E. coli (wild-type)
  • Enzyme purity: >99%
  • Required cofactor: ATP
  • Biotinylation type: Covalent
  • Standard kit enzyme concentration: 1 mg/mL
  • Standard kit enzyme amount: 40 μg
  • Main application: Protein labeling and detection
  • Key benefit: High efficiency and specificity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.