Learn More
Medchemexpress LLC BirA protein, E. coli (His-MBP) | >95.0% | 10 UG

Supplier: Medchemexpress LLC HYP7547510UG
Recombinant biotin ligase (BirA) derived from Escherichia coli, expressed in E. coli and supplied as an N-terminal 6xHis-MBP fusion protein. The product is lyophilized with stabilizers and intended for research use; biological activity is measured by pyrophosphate generation in the biotinylation reaction.
- Expression host: Escherichia coli.
- Tags: N-terminal 6xHis and MBP.
- Protein length: 321 amino acids, full length.
- Purity: >95% by reducing SDS-PAGE.
- Formulation: lyophilized from Tris/NaCl with glycerol, trehalose, mannitol, and Tween-80.
- Activity assay: biotinylation measured via pyrophosphate generation.
- Storage and handling: store at -20°C; reconstitute to ≥100 μg/mL in ddH2O recommended.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.