Learn More
Medchemexpress LLC BirA protein, E. coli (His-MBP) | >95.0% | 5 UG
SDP
Catalog No. 5000291330
Encompass_Preferred
Recombinant BirA protein from Escherichia coli, expressed in E. coli and produced with N-terminal 6xHis and maltose-binding protein (MBP) fusion tags. Supplied as a purified enzyme for in vitro biotinylation and related biochemical applications, the product is offered in multiple microgram pack sizes and includes batch-specific certificates of analysis.
Catalog No. 50-002-91330 Supplier Medchemexpress LLC Supplier No. HYP754755UG
Recombinant BirA protein from Escherichia coli, expressed in E. coli and produced with N-terminal 6xHis and maltose-binding protein (MBP) fusion tags. Supplied as a purified enzyme for in vitro biotinylation and related biochemical applications, the product is offered in multiple microgram pack sizes and includes batch-specific certificates of analysis.