Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVIDITY LLC BIRA500-RT Biotin Protein Ligase, 2 vials of 20 μL each, 40 μg total, Lyophilized reaction kit, Covalent addition of d-biotin via ATP, >99% purity

Catalog No. NC2047451
Encompass

The BIRA500-RT Biotin Protein Ligase is a lyophilized reaction kit designed for efficient and targeted biotinylation. This process is crucial in various applications such as affinity chromatography, protein immobilization, and detection with anti-biotin antibodies or avidin conjugates. Optimized for use with AviTag™ peptides, the kit enhances reaction rates, reduces enzyme use, and shortens incubation times, minimizing issues related to proteases and protein instability.

  • Enzyme name: BirA biotin-protein ligase
  • EC number: 6.3.4.15
  • Form: Lyophilized reaction kit
  • Source: E. coli wild-type
  • Purity: >99%
  • Molecular weight: 33,500 Daltons
  • Activity: >8000 Units/μg of BirA
  • Recommended substrate: AviTag™ peptide sequences
  • Stored lyophilized: 1 year at 4°C
  • Reconstituted storage: 2 weeks at 4°C, long term at -80°C
  • Price: $250.00

Catalog No. NC2047451 Supplier AVIDITY LLC Supplier No. BIRA500RT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The BIRA500-RT Biotin Protein Ligase is a lyophilized reaction kit designed for efficient and targeted biotinylation. This process is crucial in various applications such as affinity chromatography, protein immobilization, and detection with anti-biotin antibodies or avidin conjugates. Optimized for use with AviTag™ peptides, the kit enhances reaction rates, reduces enzyme use, and shortens incubation times, minimizing issues related to proteases and protein instability.

  • Enzyme name: BirA biotin-protein ligase
  • EC number: 6.3.4.15
  • Form: Lyophilized reaction kit
  • Source: E. coli wild-type
  • Purity: >99%
  • Molecular weight: 33,500 Daltons
  • Activity: >8000 Units/μg of BirA
  • Recommended substrate: AviTag™ peptide sequences
  • Stored lyophilized: 1 year at 4°C
  • Reconstituted storage: 2 weeks at 4°C, long term at -80°C
  • Price: $250.00

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.