Learn More
Apexbio Technology LLC BIRB 796 (Doramapimod), 10mg. Cas: 285983-48-4 MFCD: MFCD09752957. P38 MAPK inhibitor,cell permeable and highly selective
SDP
Supplier:  Apexbio Technology LLC A563910
Encompass_Preferred
EC50: 18 nM for TNF- in THP-1 cellsThe signal transduction pathway leading to the production of TNF- from stimulated inflammatory cells, while not fully understood, has been shown to be partially regulated by p38 mitogen activated protein (MAP) kinase. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4959