Learn More
Apexbio Technology LLC BIRB 796 (Doramapimod) 285983-48-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062439
Encompass_Preferred
BIRB 796 (Doramapimod CAS 285983-48-4) is a potent inhibitor of human p38 mitogen-activated protein kinase (MAPK) which modulates the signaling pathways associated with inflammatory cytokine production It demonstrates significant inhibition of TNF- release (EC50 18 nM) in THP-1 cells In animal models oral administration reduces LPS-induced TNF- synthesis and ameliorates collagen-induced arthritis severity Clinically however trials for Crohn s disease showed no superiority over placebo BIRB 796 serves as a valuable research tool in inflammation and cytokine signaling studies
Catalog No. 50-000-62439 Supplier Apexbio Technology LLC Supplier No. A56395.1
BIRB 796 (Doramapimod CAS 285983-48-4) is a potent inhibitor of human p38 mitogen-activated protein kinase (MAPK) which modulates the signaling pathways associated with inflammatory cytokine production It demonstrates significant inhibition of TNF- release (EC50 18 nM) in THP-1 cells In animal models oral administration reduces LPS-induced TNF- synthesis and ameliorates collagen-induced arthritis severity Clinically however trials for Crohn s disease showed no superiority over placebo BIRB 796 serves as a valuable research tool in inflammation and cytokine signaling studies