Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Baculoviral IAP repeat-containing protein 5 (survivin) | >95.0% | 100UG
SDP

Catalog No. 5000175094
Encompass_Preferred

Recombinant full-length human BIRC5 (survivin) with N-terminal 6xHis and SUMO fusion tags, expressed in E. coli and supplied lyophilized for research applications. The protein is intended for activity and cell proliferation assays and includes manufacturer guidance for storage and handling.

  • Full-length human isoform-1 sequence.
  • N-terminal 6xHis and SUMO fusion tags for purification.
  • Expressed in E. coli and purified to >95.0% as determined by reducing SDS-PAGE.
  • Lyophilized from PBS, pH 7.4, and supplied as measured mass vials (5 μg, 10 μg, 50 μg, 100 μg).
  • Activity demonstrated in A549 cell proliferation assay (ED50 0.022 μg/mL).
  • Recommended storage at -20°C; reconstituted material stable at 4°C for 1 week or frozen for long-term storage.

Catalog No. 50-001-75094 Supplier Medchemexpress LLC Supplier No. HYP702495100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human BIRC5 (survivin) with N-terminal 6xHis and SUMO fusion tags, expressed in E. coli and supplied lyophilized for research applications. The protein is intended for activity and cell proliferation assays and includes manufacturer guidance for storage and handling.

  • Full-length human isoform-1 sequence.
  • N-terminal 6xHis and SUMO fusion tags for purification.
  • Expressed in E. coli and purified to >95.0% as determined by reducing SDS-PAGE.
  • Lyophilized from PBS, pH 7.4, and supplied as measured mass vials (5 μg, 10 μg, 50 μg, 100 μg).
  • Activity demonstrated in A549 cell proliferation assay (ED50 0.022 μg/mL).
  • Recommended storage at -20°C; reconstituted material stable at 4°C for 1 week or frozen for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.