Learn More
Medchemexpress LLC BIRC5 protein, human (His-SUMO) | >95.0% | 10UG
SDP
Catalog No. 5000176632
Encompass_Preferred
Recombinant human BIRC5 (Survivin) with an N-terminal 6xHis-SUMO fusion tag, expressed in E. coli and supplied lyophilized for biochemical and cell biology research. The purified protein (≈35 kDa, 142 aa) is >95% pure by reducing SDS-PAGE and includes formulation and storage guidance for reliable assay performance.
  • Recombinant human BIRC5 with N-terminal His-SUMO tag
  • Expressed in E. coli and purified to high purity (>95%)
  • Approximate molecular weight 35 kDa and sequence length 142 amino acids
  • Lyophilized formulation for stable storage and convenient reconstitution
  • Suitable for antibody validation, biochemical assays, and functional studies
Catalog No. 50-001-76632 Supplier Medchemexpress LLC Supplier No. HYP70249510UG
Recombinant human BIRC5 (Survivin) with an N-terminal 6xHis-SUMO fusion tag, expressed in E. coli and supplied lyophilized for biochemical and cell biology research. The purified protein (≈35 kDa, 142 aa) is >95% pure by reducing SDS-PAGE and includes formulation and storage guidance for reliable assay performance.
  • Recombinant human BIRC5 with N-terminal His-SUMO tag
  • Expressed in E. coli and purified to high purity (>95%)
  • Approximate molecular weight 35 kDa and sequence length 142 amino acids
  • Lyophilized formulation for stable storage and convenient reconstitution
  • Suitable for antibody validation, biochemical assays, and functional studies