Learn More
Med Vet International Bird Nail & Feather Trimmer, 10cm L, J0391
Catalog No. 5000585352
Encompass_Preferred
The very small nails of birds require delicate trimming to avoid bleeding or injury. Other trimmers are simply too large to do a proper job. Stainless steel notched blades.
Catalog No. 50-005-85352 Supplier Med Vet International Supplier No. JOR391
The very small nails of birds require delicate trimming to avoid bleeding or injury. Other trimmers are simply too large to do a proper job. Stainless steel notched blades.