Learn More
MSC Bird-X Sonic Bird Deterrent, 1 Acre Coverage, Weather Resistant, Programmable
Catalog No. 502835841
Encompass
This sonic bird deterrent uses naturally recorded distress calls and predator cries to safely and humanely scare away pest birds. The device is fully programmable, providing clear and high-definition playback for effective bird control in various indoor and outdoor settings.
  • Deters pest birds without traps or chemicals.
  • Uses naturally recorded bird calls and predator cries.
  • Covers up to 1 acre of land.
  • Completely weather resistant for outdoor use.
  • Fully programmable sound, volume, and timing options.
  • Harmless, using natural sounds to chase away birds.
  • Compatible with a small solar panel and battery.
  • Reduces health and liability risks.
  • Maintains property aesthetics.
  • Installs easily with no additional hardware required.
  • Photocell sensor allows programming to turn on at dawn.
Catalog No. 50-283-5841 Supplier MSC Supplier No. 72510894
This sonic bird deterrent uses naturally recorded distress calls and predator cries to safely and humanely scare away pest birds. The device is fully programmable, providing clear and high-definition playback for effective bird control in various indoor and outdoor settings.
  • Deters pest birds without traps or chemicals.
  • Uses naturally recorded bird calls and predator cries.
  • Covers up to 1 acre of land.
  • Completely weather resistant for outdoor use.
  • Fully programmable sound, volume, and timing options.
  • Harmless, using natural sounds to chase away birds.
  • Compatible with a small solar panel and battery.
  • Reduces health and liability risks.
  • Maintains property aesthetics.
  • Installs easily with no additional hardware required.
  • Photocell sensor allows programming to turn on at dawn.