Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) | 31277-98-2 | MFCD00009880 | 98.0% | 903.25 g·mol-1 | C52H48P4Pd | 5 G
SDP

Catalog No. 5000300887
Encompass_Preferred

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) is a palladium(0) complex intended for research use as a reagent and catalyst precursor in laboratory synthesis and biochemical assays. It is supplied as a light yellow to brown solid with documented composition and purity for reproducible experimental results.

  • High purity (≈98.0%) for reliable experimental performance.
  • Chemical formula C52H48P4Pd and molecular weight 903.25 g·mol-1 for accurate stoichiometry.
  • Available in lab-scale packaging appropriate for bench synthesis.
  • Storage stability when kept under inert atmosphere at -20°C.
  • Datasheet, COA, and SDS available to support safe handling and quality verification.

Catalog No. 50-003-00887 Supplier Medchemexpress LLC Supplier No. HYW0209855G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) is a palladium(0) complex intended for research use as a reagent and catalyst precursor in laboratory synthesis and biochemical assays. It is supplied as a light yellow to brown solid with documented composition and purity for reproducible experimental results.

  • High purity (≈98.0%) for reliable experimental performance.
  • Chemical formula C52H48P4Pd and molecular weight 903.25 g·mol-1 for accurate stoichiometry.
  • Available in lab-scale packaging appropriate for bench synthesis.
  • Storage stability when kept under inert atmosphere at -20°C.
  • Datasheet, COA, and SDS available to support safe handling and quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.