Learn More
Medchemexpress LLC Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) | 31277-98-2 | MFCD00009880 | ≥98.0% | 903.25 g·mol⁻¹ | C52H48P4Pd | 1 G
SDP
Catalog No. 5000301988
Encompass_Preferred
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) is a zero-valent palladium complex supplied as a light yellow to brown solid. It is used as a preformed Pd(0) source for homogeneous catalysis in cross-coupling and related organic transformations, providing consistent activity without in-situ reduction steps.
  • Zero-valent palladium complex for homogeneous catalysis.
  • Suitable as a preformed Pd(0) source for cross-coupling reactions.
  • High purity (≥98.0%) for reproducible performance.
  • Molecular weight 903.25 g·mol⁻¹ and formula C52H48P4Pd.
  • Air-sensitive; store at -20 °C under inert atmosphere.
Catalog No. 50-003-01988 Supplier Medchemexpress LLC Supplier No. HYW0209851G
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) is a zero-valent palladium complex supplied as a light yellow to brown solid. It is used as a preformed Pd(0) source for homogeneous catalysis in cross-coupling and related organic transformations, providing consistent activity without in-situ reduction steps.
  • Zero-valent palladium complex for homogeneous catalysis.
  • Suitable as a preformed Pd(0) source for cross-coupling reactions.
  • High purity (≥98.0%) for reproducible performance.
  • Molecular weight 903.25 g·mol⁻¹ and formula C52H48P4Pd.
  • Air-sensitive; store at -20 °C under inert atmosphere.