Learn More
Medchemexpress LLC Bis(1,3-dichloro-2-propyl) phosphate-d10 | 1477495-19-4 | ≥99.0% | C6HD10Cl4O4P | 1 MG
SDP
Catalog No. 5000353754
Encompass_Preferred
Bis(1,3-dichloro-2-propyl) phosphate-d10 is the deuterium labeled Bis(1,3-dichloro-2-propyl) phosphate. It is intended for research use and not for patient administration. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-53754 Supplier Medchemexpress LLC Supplier No. HY141771S1MG
Bis(1,3-dichloro-2-propyl) phosphate-d10 is the deuterium labeled Bis(1,3-dichloro-2-propyl) phosphate. It is intended for research use and not for patient administration. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.