Learn More
Strem, An Ascensus Company CAS# 35138-23-9. Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate, 95%. MFCD00079664. 500MG
SDP
Supplier:  Strem, An Ascensus Company 770200500MG
Encompass
CAS# 35138-23-9. 500MG. Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate, 95%. MFCD00079664. Molecular Formula: C16H24BF4Ir Molecular Weight: 495.40. Color/Form: dark red xtl. Stability: air sensitive, store cold Strem#: 77-0200 www.strem.com/catalog/v3/77-0200/
Catalog No. 50-901-13100