Learn More
Strem, An Ascensus Company CAS# 1295-35-8. 10g. Bis(1,5-cyclooctadiene)nickel (0), 98+%. MFCD00058902
SDP
Supplier:  Strem, An Ascensus Company 28001010G
Encompass
CAS# 1295-35-8. 10g. Bis(1,5-cyclooctadiene)nickel (0), 98+%. MFCD00058902. Molecular Weight: 275.08. Molecular Formula: (C8H12)2Ni. Color/form: yellow xtl. Strem# 28-0010. http://www.strem.com/catalog/v/28-0010/
Catalog No. 50-901-11215