Alfa Aesar™ Bis(1,5-cyclooctadiene)nickel(0), 96%

Manufacturer: Alfa Aesar™ 4415704

Quantity 2g

 View more versions of this product

Catalog No. AA4415704

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1295-35-8
    Molecular Formula C16H24Ni
    Formula Weight 275.06
    MDL Number MFCD00058902
    UN Number UN1325
    Chemical Name or Material Bis (1,5-cyclooctadiene)nickel (0)

    Specifications

    Formula Weight 275.06
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C16H24Ni
    CAS 1295-35-8
    Melting Point 60°C (dec.)
    Storage Note 1
    • Packaged under argon
    Physical Form Powder
    MDL Number MFCD00058902
    UN Number UN1325
    Quantity 2g
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Odor Odorless
    Color Yellow
    Sensitivity Air & Light Sensitive
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H350-H317
    Flammable solid.
    May cause cancer.
    May cause an allergic skin reaction.
    RTECS Number QR6135000
    Chemical Name or Material Bis (1,5-cyclooctadiene)nickel (0)
    Assay 96%

    Description

    • Catalyst for isomerization and hydrosilyltion of unsaturated compounds
    • Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene

    GHS H Statement
    H228-H350-H317
    Flammable solid.
    May cause cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit