Learn More
Strem, An Ascensus Company Bis(COD)Rh(I) BF4, 98+% 100mg
SDP
Catalog No. 5090112051
Encompass
CAS# 35138-22-8. 100mg. Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+%. MFCD00075045. Molecular Weight: 406.07. Molecular Formula: Rh(C8H12)2+BF4ae. Color/form: red-brown solid Strem# 45-0109. http://www.strem.com/catalog/v/45-0109/
Catalog No. 50-901-12051 Supplier Strem, An Ascensus Company Supplier No. 450109100MG
CAS# 35138-22-8. 100mg. Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+%. MFCD00075045. Molecular Weight: 406.07. Molecular Formula: Rh(C8H12)2+BF4ae. Color/form: red-brown solid Strem# 45-0109. http://www.strem.com/catalog/v/45-0109/