Learn More
Strem, An Ascensus Company Bis(COD)rhodium(I) BF4,98+% 2g
SDP
Catalog No. 5090112053
Encompass
CAS# 35138-22-8. 2g. Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+%. MFCD00075045. Molecular Weight: 406.07. Molecular Formula: Rh(C8H12)2+BF4ae. Color/form: red-brown solid Strem# 45-0109. http://www.strem.com/catalog/v/45-0109/
Catalog No. 50-901-12053 Supplier Strem, An Ascensus Company Supplier No. 4501092G
CAS# 35138-22-8. 2g. Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+%. MFCD00075045. Molecular Weight: 406.07. Molecular Formula: Rh(C8H12)2+BF4ae. Color/form: red-brown solid Strem# 45-0109. http://www.strem.com/catalog/v/45-0109/