Learn More
Medchemexpress LLC Bis(1-chloro-2-propyl) phosphate | 789440-10-4 | MFCD31579781 | 251.04 g/mol | C6H13Cl2O4P | 1 MG
SDP
Catalog No. 5000284968
Encompass_Preferred
Bis(1-chloro-2-propyl) phosphate is an organophosphate research chemical and a metabolite reference standard used in studies of organophosphate flame retardants and their metabolites. It is supplied as a small laboratory quantity for analytical and toxicological research.
  • Chemical identifiers: CAS 789440-10-4; formula C6H13Cl2O4P; molecular weight 251.04 g/mol.
  • Used as an analytical standard for metabolite identification and biomonitoring studies.
  • Provided as a 1 mg laboratory sample suitable for analytical assays and method development.
  • Intended for laboratory research use; consult the safety data sheet for handling and hazard information.
Catalog No. 50-002-84968 Supplier Medchemexpress LLC Supplier No. HYW6539291MG
Bis(1-chloro-2-propyl) phosphate is an organophosphate research chemical and a metabolite reference standard used in studies of organophosphate flame retardants and their metabolites. It is supplied as a small laboratory quantity for analytical and toxicological research.
  • Chemical identifiers: CAS 789440-10-4; formula C6H13Cl2O4P; molecular weight 251.04 g/mol.
  • Used as an analytical standard for metabolite identification and biomonitoring studies.
  • Provided as a 1 mg laboratory sample suitable for analytical assays and method development.
  • Intended for laboratory research use; consult the safety data sheet for handling and hazard information.