Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Bis[™-1-Phenylethyl]Amine

Catalog No. 45282310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-282-310G 10 g
45-282-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 45-282-310G Supplier Sigma Aldrich Supplier No. 45282310G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+)-Bis[™-1-phenylethyl]amine can be used:In the synthesis of+a-amino acids.In the preparation of chiral C(19)-C(26) and C(27)-C(32) moieties of scytophycin C.To induce enantioselectivity in the deprotonation of prochiral ketones.As a starting material for the synthesis of chiral phenolate ligands through Mannich condensation reaction.In the synthesis of chiral phosphoramidite ligands.To prepare chiral cyclic isoimidium salts, which are further used to synthesize chiral lactones through Diels-Alder reaction.

Specifications

CAS 23294-41-9
Density 0.985 g/mL at 25°C
Boiling Point 86°C (0.05 mmHg, lit.)
Molecular Formula C16H19N
Linear Formula [C6H5CH(CH3)]2NH
Refractive Index n20/D 1.5523
Quantity 10 g
MDL Number MFCD00243088
Synonym (+)-Bis[(R)-alpha-methylbenzyl]amine; [R-(R,R)]-(+)-Bis(alpha-methylbenzyl)amine
Molecular Weight (g/mol) 225.33 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.