Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,2,2-trifluoroethyl) Phthalate 98.0+%, TCI America™
SDP

Catalog No. P0785100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0785100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0785100MG Supplier TCI America Supplier No. P0785100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62240-27-1
Molecular Formula C12H8F6O4
Molecular Weight (g/mol) 330.18
MDL Number MFCD00059472
InChI Key PSRBRNHUQJKQHV-UHFFFAOYSA-N
Synonym Phthalic Acid Bis(2,2,2-trifluoroethyl) Ester
PubChem CID 615877
IUPAC Name 1,2-bis(2,2,2-trifluoroethyl) benzene-1,2-dicarboxylate
SMILES FC(F)(F)COC(=O)C1=CC=CC=C1C(=O)OCC(F)(F)F

Specifications

Melting Point 41°C
Color White
Quantity 100 mg
Formula Weight 330.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,2,2-trifluoroethyl) Phthalate [Standard for Phthalate GLC Determination]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.