Learn More
Strem, An Ascensus Company CAS# 14481-08-4. 1g. Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), min. 98% (99.9%-Ni) [Ni(TMHD)2]. MFCD00192348
SDP
Supplier:  Strem, An Ascensus Company 2800881G
Encompass
CAS# 14481-08-4. 1g. Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), min. 98% (99.9%-Ni) [Ni(TMHD)2]. MFCD00192348. Molecular Weight: 425.23. Molecular Formula: Ni(C11H19O2)2. Color/form: purple xtl. Strem# 28-0088. http://www.strem.com/catalog/v/28-0088/
Catalog No. 50-901-11229