Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II), 99.99% (metals basis)

Catalog No. AA4158902
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4158902 0.5 g
AA4158904 2 g
AA4158909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4158902 Supplier Thermo Scientific Chemicals Supplier No. 4158902
Add to Cart

CAS: 118448-18-3 | C22H38CaO4 | 406.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118448-18-3
Molecular Formula C22H38CaO4
Molecular Weight (g/mol) 406.62
MDL Number MFCD00064757
InChI Key DOOFPPIHJGRIGW-OLHSYEKISA-L
Synonym ca tmhd 2, calcium bis dipivaloylmethanate, bis 2,2,6,6-tetramethyl-3,5-heptanedionato calcium
PubChem CID 14598209
IUPAC Name calcium;(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;(E)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Ca+2]

Specifications

Melting Point 202°C to 206°C
Boiling Point 205°C (0.1 mmHg)(sublimation)
Quantity 0.5 g
Sensitivity Hygroscopic
Formula Weight 406.57
Percent Purity 99.99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.