Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis[2-(2-bromoisobutyryloxy)undecyl] disulfide

Catalog No. 733350500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-335-0500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 73-335-0500MG Supplier Sigma Aldrich Supplier No. 733350500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Atom transfer Radical Polymerisation (ATRP) initiator commonly used to functionalize noble metal surfaces, and in the preparation of polymer brushes and biodegradable hydrogels. Also may be used to introduce a temperature and light sensitive cleavable region into the polymer.

Specifications

CAS 402828-41-5
Density 1.177 g/mL (at 25°C)
Molecular Formula C30H56Br2O4S2
Linear Formula C30H56Br2O4S2
Quantity 500 mg
Synonym 11,11 -Dithiobis[1-(2-bromo-2-methylpropionyloxy)undecane]; DTBU
Molecular Weight (g/mol) 704.7
Percent Purity 97%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.