Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis[2-(2-chloroethoxy)ethyl] ether

Catalog No. 1462550ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-625-50ML 50 mL
14-625-10ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-625-50ML Supplier Sigma Aldrich Supplier No. 1462550ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Bis[2-(2-chloroethoxy)ethyl] ether was used in preparation of series of para-(1,1,3,3-tetramethylbutyl)-phenoxypoly(ethoxy)ethanols
  • It was used in the synthesis of 1-(4-pyridiniumaldoxime)-2-(((2-chloroethoxy)-2-ethoxy)ethoxy)ethane

Specifications

CAS 638-56-2
Density 1.18 g/mL (at 20°C (literature))
Molecular Formula C8H16Cl2O3
Refractive Index n20/D 1.464
Linear Formula O(CH2CH2OCH2CH2Cl)2
Quantity 50 mL
MDL Number MFCD00054473
Synonym 1,11-Dichloro-3,6,9-trioxaundecane; Tetraethylene glycol dichloride
Molecular Weight (g/mol) 231.12
Percent Purity ≥99.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.