Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,4,5-trichlorophenyl) Disulfide 95.0+%, TCI America™
SDP

Catalog No. B024525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B024525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B024525G Supplier TCI America Supplier No. B024525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3808-87-5
Molecular Formula C12H4Cl6S2
Molecular Weight (g/mol) 424.98
MDL Number MFCD00000551
InChI Key ZUVJVEOHQKNMPN-UHFFFAOYSA-N
Synonym bis 2,4,5-trichlorophenyl disulfide, 2,4,5-trichlorophenyl disulfide, unii-ee043utu8o, 1,2,4-trichloro-5-2,4,5-trichlorophenyl disulfanyl benzene, 2,4,5-trichlorophenyl disulphide, ee043utu8o, bis 2,4,5-trichlorophenyl disulphide, 1,1'-dithiobis 2,4,5-trichlorobenzene, acmc-1ajg6
PubChem CID 77421
IUPAC Name 1,2,4-trichloro-5-[(2,4,5-trichlorophenyl)disulfanyl]benzene
SMILES ClC1=CC(Cl)=C(SSC2=CC(Cl)=C(Cl)C=C2Cl)C=C1Cl

Specifications

Melting Point 147°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 424.98
Percent Purity ≥95.0% (GC)
Chemical Name or Material Bis(2,4,5-trichlorophenyl) Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.