Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(2,4,6-trichlorophenyl) oxalate | 1165-91-9 | MFCD00043061 | 98.0% | 448.90 g/mol | C14H4Cl6O4 | 100 G
SDP

Catalog No. 5000210058
Encompass_Preferred

Bis(2,4,6-trichlorophenyl) oxalate (CAS 1165-91-9) is an organic oxalate reagent supplied as a solid for use in biochemical assays and peroxyoxalate chemiluminescence systems. It is provided at high purity for laboratory-scale applications.

  • High purity (98.0%).
  • Molecular formula C14H4Cl6O4 and molecular weight 448.90 g/mol.
  • White solid physical form suitable for handling in analytical and synthetic workflows.
  • Used in peroxyoxalate chemiluminescence and biochemical assay applications.
  • Available in laboratory pack sizes, including 100 G.
  • Recommended storage: store solid at room temperature; in solvent, follow cold storage guidance.

Catalog No. 50-002-10058 Supplier Medchemexpress LLC Supplier No. HYW042438100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis(2,4,6-trichlorophenyl) oxalate (CAS 1165-91-9) is an organic oxalate reagent supplied as a solid for use in biochemical assays and peroxyoxalate chemiluminescence systems. It is provided at high purity for laboratory-scale applications.

  • High purity (98.0%).
  • Molecular formula C14H4Cl6O4 and molecular weight 448.90 g/mol.
  • White solid physical form suitable for handling in analytical and synthetic workflows.
  • Used in peroxyoxalate chemiluminescence and biochemical assay applications.
  • Available in laboratory pack sizes, including 100 G.
  • Recommended storage: store solid at room temperature; in solvent, follow cold storage guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.