Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[2-(4-azidosalicylamido)ethyl] Disulfide 97.0+%, TCI America™
SDP

Catalog No. B379010MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B379010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B379010MG Supplier TCI America Supplier No. B379010MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 199804-21-2
Molecular Formula C18H18N8O4S2
Molecular Weight (g/mol) 474.514
MDL Number MFCD01861964
InChI Key QQZOUYFHWKTGEY-UHFFFAOYSA-N
Synonym BASED
PubChem CID 3658531
IUPAC Name 4-azido-N-[2-[2-[(4-azido-2-hydroxybenzoyl)amino]ethyldisulfanyl]ethyl]-2-hydroxybenzamide
SMILES C1=CC(=C(C=C1N=[N+]=[N-])O)C(=O)NCCSSCCNC(=O)C2=C(C=C(C=C2)N=[N+]=[N-])O

Specifications

Quantity 10 mg
Formula Weight 474.51
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis[2-(4-azidosalicylamido)ethyl] Disulfide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.