Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) (75% in Isopropyl Alcohol), TCI America™
SDP

Catalog No. B339525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B339525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B339525G Supplier TCI America Supplier No. B339525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17927-72-9
Molecular Formula C16H28O6Ti
MDL Number MFCD00000031
Synonym Titanium(IV) Diisopropoxide Bis(acetylacetonate), Diisopropoxytitanium(IV) Bis(acetylacetonate)

Specifications

Quantity 25 g
UN Number 1219
Formula Weight 364.26
Physical Form Liquid
Chemical Name or Material Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) (75% in Isopropyl Alcohol)
EINECSNumber (2)-2161
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.