Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,4-pentanedionato)cobalt(II) 97.0+%, TCI America™
SDP

Catalog No. B268125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B268125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B268125G Supplier TCI America Supplier No. B268125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14024-48-7
Molecular Formula C10H14CoO4
Molecular Weight (g/mol) 257.15
MDL Number MFCD00000014
InChI Key UTYYEGLZLFAFDI-FDGPNNRMSA-L
Synonym Acetylacetone Cobalt(II) Salt, Cobalt(II) Acetylacetonate
IUPAC Name λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 173°C
Quantity 25 g
Formula Weight 257.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4-pentanedionato)cobalt(II)
EINECSNumber (2)-2142&(2)-2181
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.