Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,4-pentanedionato)tin(IV) Dichloride 98.0+%, TCI America™
SDP

Catalog No. T065025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T065025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T065025G Supplier TCI America Supplier No. T065025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16919-46-3
Molecular Formula C10H14Cl2O4Sn
Molecular Weight (g/mol) 387.83
MDL Number MFCD00015316
InChI Key DTMARTPDQMQTRX-VGKOASNMSA-J
Synonym Acetylacetone Tin(IV) Dichloride Salt, Tin(IV) Acetylacetonate Dichloride
IUPAC Name tin(4+) bis((2Z)-4-oxopent-2-en-2-olate) dichloride
SMILES [Cl-].[Cl-].[Sn+4].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 206°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3146
Formula Weight 387.83
Percent Purity ≥98.0% (T)
Chemical Name or Material Bis(2,4-pentanedionato)tin(IV) Dichloride
EINECSNumber (2)-2246
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.