Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,4-pentanedionato)titanium(IV) Oxide, TCI America™
SDP

Catalog No. T024925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T024925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T024925G Supplier TCI America Supplier No. T024925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14024-64-7
Molecular Formula C10H16O5Ti
Molecular Weight (g/mol) 264.1
MDL Number MFCD00013505
InChI Key ADVORQMAWLEPOI-UHFFFAOYSA-N
Synonym bis acetylacetonato titanium iv oxide, bis 4-hydroxypent-3-en-2-one ; titanium ii oxide
PubChem CID 74765597
IUPAC Name 4-hydroxypent-3-en-2-one;oxotitanium
SMILES CC(=CC(=O)C)O.CC(=CC(=O)C)O.O=[Ti]

Specifications

Melting Point 200°C
Quantity 25 g
Formula Weight 262.08
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4-pentanedionato)titanium(IV) Oxide
EINECSNumber (2)-2142&(2)-2994
RTECSNumber XR2330000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.