Learn More
Medchemexpress LLC Bis(2-bromoethyl) ether | 5414-19-7 | 99.77% | 231.92 | 500 G
SDP
Catalog No. 5000454743
Encompass_Preferred
Bis(2-bromoethyl) ether is an alkyl chain-based PROTAC linker. It can be utilized in the synthesis of PROTACs, which leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs.
  • Functions by leveraging the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
Catalog No. 50-004-54743 Supplier Medchemexpress LLC Supplier No. HYW013458500G
Bis(2-bromoethyl) ether is an alkyl chain-based PROTAC linker. It can be utilized in the synthesis of PROTACs, which leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs.
  • Functions by leveraging the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.