Learn More
Medchemexpress LLC Bis(2-bromoethyl) ether | 5414-19-7 | 99.8% | 231.92 | 25 G
SDP
Catalog No. 5000451400
Encompass_Preferred
Bis(2-bromoethyl) ether is an alkyl chain-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs themselves consist of two distinct ligands joined by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These molecules function by harnessing the intracellular ubiquitin-proteasome system to achieve the selective degradation of target proteins.
  • It is an alkyl chain-based linker.
  • It can be used in the synthesis of PROTACs.
  • The linked PROTACs exploit the intracellular ubiquitin-proteasome system.
  • The linked PROTACs selectively degrade target proteins.
Catalog No. 50-004-51400 Supplier Medchemexpress LLC Supplier No. 25GHYW013458
Bis(2-bromoethyl) ether is an alkyl chain-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs themselves consist of two distinct ligands joined by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These molecules function by harnessing the intracellular ubiquitin-proteasome system to achieve the selective degradation of target proteins.
  • It is an alkyl chain-based linker.
  • It can be used in the synthesis of PROTACs.
  • The linked PROTACs exploit the intracellular ubiquitin-proteasome system.
  • The linked PROTACs selectively degrade target proteins.