Learn More
Medchemexpress LLC Bis(2-bromoethyl) ether | 5414-19-7 | 99.9% | 231.92 | 10 G
SDP
Catalog No. 5000454236
Encompass_Preferred
Bis(2-bromoethyl) ether is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs (Proteolysis Targeting Chimeras). PROTACs are designed to degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • Alkyl chain-based PROTAC linker used in the synthesis of PROTACs.
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system.
Catalog No. 50-004-54236 Supplier Medchemexpress LLC Supplier No. HYW01345810G
Bis(2-bromoethyl) ether is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs (Proteolysis Targeting Chimeras). PROTACs are designed to degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • Alkyl chain-based PROTAC linker used in the synthesis of PROTACs.
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system.